Anti-CD8A Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 53-6.7]
USBIC2259-11D
- Antibody type:Primary
- Antigen name:CD8a molecule
- Antigen symbol:CD8A
- Clonality:Monoclonal
- Clone:53-6.7
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Form:liquid
- Storage buffer:50mMsodium Phosphate pH 7,5, 500mM Potassium chloride, 150mM NaCl, 15% glycerol, 0,2% BSA, 0,04% NaN3.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:120 Tests
- Pk:120 Tests
Do not freeze!
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse