Anti-CD8A Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 53-6.7]
Catalog # USBIC2259-11C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:CD8a molecule
- Antigen symbol:CD8A
- Clonality:Monoclonal
- Clone:53-6.7
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Form:liquid
- Storage buffer:50mMsodium phosphate pH 7,5, 100mM potassium chloride, 150mM sodium chloride, 5% glycerol, 0,2% BSA, 0,04% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:120 Tests
- Pk:120 Tests
Specifications
About this item
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse