Anti-CD8A Rat Monoclonal Antibody (Biotin) [clone: 53-6.7]
USBIC2259-11B
- Antibody type:Primary
- Antigen name:CD8a molecule
- Antigen symbol:CD8A
- Clonality:Monoclonal
- Clone:53-6.7
- Conjugation:Biotin
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Form:liquid
- Storage buffer:FBS,50mM sodium Phosphate pH 7,5, 100mM Potassium chloride, 150mM NaCl, 0,5 mg/ml GentamicSulfate.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse