Anti-CD8A Mouse Monoclonal Antibody [clone: 15-11C5]
USBIC2259-03E
- Antibody type:Primary
- Antigen name:CD8a molecule
- Antigen symbol:CD8A
- Clonality:Monoclonal
- Clone:15-11C5
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, 0,01% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:1 ml
- Pk:1 mL
Recommended dilution: Flow Cytometry: 1:10; Immunochemistry: 1:100
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: Monoclonal
Clone: 15-11C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human