Anti-CEA Mouse monoclonal antibody unconjugated [clone: CLB-139]
USBIC1300-01M
- Antibody type:Primary
- Antigen name:Carcinoembryonic Antigen
- Antigen symbol:CEA
- Clonality:Monoclonal
- Clone:CLB-139
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:20mMTRIS, 150mM sodium chloride, pH 8.0, 10mg BSA, 0,001% mertiolate.
- Storage temperature:Cool
- Concentration:~0,2 mg/ml
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:1 ml
- Pk:1 mL
Type: Primary
Antigen: CEA (Carcino Embryonic Antigen)
Clonality: Monoclonal
Clone: CLB-139
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human