Anti-CASQ2 Mouse Monoclonal Antibody [clone: 1B6]
Catalog # USBIC1036-07
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:calsequestrin 2 (cardiac muscle)
- Antigen symbol:CASQ2
- Clonality:Monoclonal
- Clone:1B6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CASQ2 (calsequestrin 2 (cardiac muscle))
Clonality: Monoclonal
Clone: 1B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human