Anti-CDH11 Goat Polyclonal Antibody
Catalog # USBIC0107-09A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:cadherin 11, type 2, OB-cadherin (osteoblast)
- Antigen symbol:CDH11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Reactivity:Human
- Form:liquid
- Amino acid number:26 to 37
- Storage buffer:trissaline, 0,02% sodium azide, pH 7,3, 0,5% BSA.
- Sequence:C-ERRGHLRPSFHG
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:2000; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: CDH11 (cadherin 11, type 2, OB-cadherin (osteoblast))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human