Anti-C-10 Rat Monoclonal Antibody [clone: 11n17]
Catalog # USBIC0005-09A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:C-10
- Clonality:Monoclonal
- Clone:11n17
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Form:lyophilized powder
- Amino acid number:22 to 116
- Storage buffer:PBS, pH 7,4, 50ug BSA
- Storage temperature:Cool
- Concentration:~0,5 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:250 µg
- Pk:250 µG
Specifications
About this item
Recommended dilution: ELISA: 2-8?g/ml; ELISA: 0.5-2?g/ml
Type: Primary
Antigen: C-10
Clonality: Monoclonal
Clone: 11n17
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse