Anti-Bu-1a Mouse Monoclonal Antibody (Biotin) [clone: 5K93]
USBIB2788-50B
- Antibody type:Primary
- Antigen symbol:BU-1A
- Clonality:Monoclonal
- Clone:5K93
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Chicken
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:500 µg
- Pk:500 µG
Type: Primary
Antigen: Bu-1a
Clonality: monoclonal
Clone: 5K93
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Chicken