Order Entry
Export
ContactUsLinkComponent
Anti-BRCC-36 Mouse monoclonal antibody unconjugated [clone: 10B2564]
Anti-BRCC-36 Mouse monoclonal antibody unconjugated [clone: 10B2564]
Catalog # USBIB2727-01C
Supplier:  US Biological
Anti-BRCC-36 Mouse monoclonal antibody unconjugated [clone: 10B2564]
Catalog # USBIB2727-01C
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    BRCC-36
  • Clonality:
    Monoclonal
  • Clone:
    10B2564
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Amino acid number:
    1 to 316
  • Storage buffer:
    PBS, pH 7,4, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: BRCC-36
Clonality: monoclonal
Clone: 10B2564
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human