Anti-BRCA1 Sheep polyclonal antibody unconjugated
Catalog # USBIB2709-06B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:breast cancer 1, early onset
- Antigen symbol:BRCA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Isotype:IgG
- Reactivity:Human
- Form:lyophilized powder
- Amino acid number:1634 to 1863
- Storage buffer:PBS,trehalose.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 5-15 ?g/ml
Type: Primary
Antigen: BRCA1 (breast cancer 1, early onset)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human