Anti-Brain Natriuretic Peptide 32 Rabbit polyclonal antibody unconjugated
Catalog # USBIB2702-14E
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Brain Natriuretic Peptide 32
- Antigen symbol:BNP32
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Pig
- Format:Serum
- Form:lyophilized powder
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Recommended dilution: Immunofluorescence: 1:200; Immunochemistry: 1:500
Type: Primary
Antigen: Brain Natriuretic Peptide 32
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pig