Anti-BNC1 Rabbit Polyclonal Antibody
USBIB0132-05
- Antibody type:Primary
- Antigen name:basonuclin 1
- Antigen symbol:BNC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human
- Western blot:Yes
- Form:liquid
- Amino acid number:330 to 380
- Storage buffer:PBS, 0,05% BSA, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 4-7?g/ml
Type: Primary
Antigen: BNC1 (basonuclin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey