Anti-BCAP31 Rabbit polyclonal antibody unconjugated
USBIB0077-04
- Antibody type:Primary
- Antigen name:B-cell receptor-associated Protein 31
- Antigen symbol:BCAP31
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
Recommended dilution: Immunochemistry: 2.5 ?g/ml; I
Type: Primary
Antigen: BCAP31 (B-cell receptor-associated protein 31)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat