Anti-BAFF Receptor Rat monoclonal antibody unconjugated [clone: 6D147]
Catalog # USBIB0003-68F
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:TNF receptor superfamily member 13C
- Antigen symbol:TNFRSF13C
- Clonality:Monoclonal
- Clone:6D147
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG1
- Reactivity:Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:2 to 70
- Storage buffer:PBS containing 0,02% sodium azide
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:100; Western Blot: 1:500
Type: Primary
Antigen: BAFF Receptor
Clonality: monoclonal
Clone: 6D147
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse