Anti-B Rabbit Polyclonal Antibody
Catalog # USBIB0002-15
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:B(QYPALT) Epitope Tag
- Antigen symbol:B(QYPALT)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:40 µg
- Pk:40 µG
Specifications
About this item
Recommended dilution: ELISA: 0. 05-0.2?g/ml; Western Blot: 0.1-1.0 ?g/ml
Type: Primary
Antigen: B
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: