Anti-NPPA Mouse Monoclonal Antibody [clone: A136-11-M14-19-H3]
Catalog # USBIA4150-03E-10
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Natriuretic peptide A
- Antigen symbol:NPPA
- Clonality:Monoclonal
- Clone:A136-11-M14-19-H3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Form:lyophilized powder
- Amino acid number:12 to 25
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:10 µg
- Pk:10 µG
Specifications
About this item
Type: Primary
Antigen: NPPA (natriuretic peptide A)
Clonality: monoclonal
Clone: A136-11-M14-19-H3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: