Order Entry
Export
ContactUsLinkComponent
Anti-Angiotensin 12 Rabbit Polyclonal Antibody
Anti-Angiotensin 12 Rabbit Polyclonal Antibody
  USBIA2295-09P
Anti-Angiotensin 12 Rabbit Polyclonal Antibody
  USBIA2295-09P

 

  • Antibody type:
    Primary
  • Antigen name:
    Angiotensin 12
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Rat
  • Format:
    Serum
  • Form:
    lyophilized powder
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: Angiotensin 12
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat