Anti-APP Mouse monoclonal antibody unconjugated [clone: 9L23]
Catalog # USBIA2275-59G
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Amyloid Beta (A4) Precursor Protein
- Antigen symbol:APP
- Clonality:Monoclonal
- Clone:9L23
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Western blot:Yes
- Form:lyophilized powder
- Antigen synonyms:ABETA|beta Amyloid
- Storage buffer:2X PBS, PEG, sucrose, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by thiophilic adsorption and size exclusion chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 0. 05 ?g/ml; Western Blot: 1 ?g/ml; Immunochemistry: 0.1-1 ?g/ml
Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: monoclonal
Clone: 9L23
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: