Order Entry
Export
ContactUsLinkComponent
Anti-Amyloid [1-28] Rabbit Polyclonal Antibody
Anti-Amyloid [1-28] Rabbit Polyclonal Antibody
Catalog # USBIA2275-58Z2
Supplier:  US Biological
Anti-Amyloid [1-28] Rabbit Polyclonal Antibody
Catalog # USBIA2275-58Z2
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Amyloid 1-28
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Format:
    Serum
  • Form:
    lyophilized powder
  • Storage buffer:
    0,1ml of 10mg/ml BSA PBS.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Tested applications:
    Radio Immunoassay
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: Amyloid [1-28]
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: