Anti-AKT1/2/3 Mouse Monoclonal Antibody (Biotin) [clone: 10g142]
Catalog # USBIA1124-12C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Protein Kinase B alpha/beta/gamma
- Antigen symbol:PKBA/PKBB/PKGG
- Clonality:Monoclonal
- Clone:10g142
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Format:ascites
- Form:liquid
- Storage buffer:136mM sodium chloride, 2.6mM KCl, 12mM sodium phosphate dibasic, pH 7,4, 2mg/ml BSA, and 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:50; Western Blot: 1:1000; Immunoprecipitation: 1:50
Type: Primary
Antigen: AKT1/2/3 (v-akt murine thymoma viral oncogene homolog 1/2/3)
Clonality: monoclonal
Clone: 10g142
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Monkey, Mouse, Rat