Anti-14-3-3 tau Goat Polyclonal Antibody
Catalog # USBI0014-20H
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:14-3-3 tau
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:Tris saline, 0,02% sodium azide, pH 7,3, 0,5% BSA.
- Sequence:C-DDRKQTIDNSQ
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:4000; Western Blot: 0. 03-0.1 ?g/ml
Type: Primary
Antigen: 14-3-3 tau (14-3-3 tau)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human, Mouse, Rat