Anti-14-3-3 eta Rabbit Polyclonal Antibody
USBI0014-06A
- Antibody type:Primary
- Antigen symbol:14-3-3 eta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:10mMsodium HEPES pH 7,5, 150mM NaCl, 0,1mg/ml BSA, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µl
- Pk:100 µl
- Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50
Type: Primary
Antigen: 14-3-3 eta (14-3-3 eta)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse, Rat