Anti-14-3-3 epsilon Rabbit Polyclonal Antibody
Catalog # USBI0014-04H
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:14-3-3 epsilon
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Sheep
- Western blot:Yes
- Epitope:N-terminal
- Format:Serum
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:3000; Immunochemistry: 1:400
Type: Primary
Antigen: 14-3-3 epsilon (14-3-3 epsilon)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Sheep