Order Entry
Export
ContactUsLinkComponent
Anti-Red Fluorescent Protein, DsRed, rDsRed Rabbit Polyclonal Antibody (Biotin)
Anti-Red Fluorescent Protein, DsRed, rDsRed Rabbit Polyclonal Antibody (Biotin)
Catalog # USBI168474
Supplier:  US Biological
Anti-Red Fluorescent Protein, DsRed, rDsRed Rabbit Polyclonal Antibody (Biotin)
Catalog # USBI168474
Supplier:  US Biological

Specifications

  • Antibody type:
    Tag
  • Antigen name:
    Red Fluorescent Protein, DsRed, rDsRed
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Form:
    lyophilized powder
  • Storage buffer:
    0,02M PBS, pH 7,2, 0,01% sodium azide, 10mg/ml BSA.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml (after reconstitution)
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: ELISA: 1:10000-1:80000; Western Blot: 1:2000-1:10000; Immunochemistry: 1:1000-1:5000

Type: Tag
Antigen: Red Fluorescent Protein, DsRed, rDsRed
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: