Anti-MIP-3a Rabbit Polyclonal Antibody
Catalog # USBI168368
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:MIP-3A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Neutralisation:Yes
- Form:lyophilized powder
- Storage buffer:0,02M PBS, pH 7,2
- Storage temperature:Cool
- Concentration:~1 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:20000-1:100000; Western Blot: 1:500-1:2000; Neutralization: 1:400
Type: Primary
Antigen: MIP-3a
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human