Anti-DbpB Rabbit Polyclonal Antibody
Catalog # USBI168141
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:DbpB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:lyophilized powder
- Storage buffer:0,02M potassium phosphate, 0,15M sodium chloride, pH 7,2, 0,01% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: DbpB
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: