Anti-SLC17A6 Mouse Monoclonal Antibody [clone: S29-29]
Catalog # USBI167618
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Solute carrier family 17 (vesicular glutamate transporter), member 6
- Antigen symbol:SLC17A6
- Clonality:Monoclonal
- Clone:S29-29
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBSpH 7,4, 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: SLC17A6 (solute carrier family 17 (vesicular glutamate transporter), member 6)
Clonality: monoclonal
Clone: S29-29
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat