Order Entry
Export
ContactUsLinkComponent
 
Anti-TARP Gamma2-4-8 Mouse Monoclonal Antibody [clone: S245-36]
  USBI167613
Anti-TARP Gamma2-4-8 Mouse Monoclonal Antibody [clone: S245-36]
  USBI167613

 

  • Antibody type:
    Primary
  • Antigen symbol:
    TARP Gamma2-4-8
  • Clonality:
    Monoclonal
  • Clone:
    S245-36
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBSpH 7,4, 50% glycerol.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: TARP Gamma2-4-8
Clonality: monoclonal
Clone: S245-36
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Rat