Anti-SYT3 Mouse Monoclonal Antibody [clone: S278-19]
USBI167609
- Antibody type:Primary
- Antigen name:Synaptotagmin 3
- Antigen symbol:SYT3
- Clonality:Monoclonal
- Clone:S278-19
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBSpH 7,4, 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: SYT3 (synaptotagmin III)
Clonality: monoclonal
Clone: S278-19
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat