Anti-ABCC8 Mouse Monoclonal Antibody [clone: S289-16]
Catalog # USBI167604
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:ATP Binding Cassette, sub-family C, member 8
- Antigen symbol:ABCC8
- Clonality:Monoclonal
- Clone:S289-16
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBSpH 7,4, 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 0.1-1 ?g/ml
Type: Primary
Antigen: ABCC8 (ATP-binding cassette, sub-family C (CFTR/MRP), member 8)
Clonality: monoclonal
Clone: S289-16
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat