Anti-mGluR1/5 glutamate receptor Mouse Monoclonal Antibody [clone: S75-33]
Catalog # USBI167598
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:mGluR1/5 Glutamate Receptor
- Antigen symbol:MGLUR1/5
- Clonality:Monoclonal
- Clone:S75-33
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBSpH 7,4, 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 0.1-1 ?g/ml
Type: Primary
Antigen: mGluR1/5 glutamate receptor
Clonality: monoclonal
Clone: S75-33
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat