Anti-GluN2A/NR2A Mouse Monoclonal Antibody [clone: S327A-38]
Catalog # USBI167592
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:GLUN2A
- Clonality:Monoclonal
- Clone:S327A-38
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Antigen synonyms:NR2A
- Storage buffer:PBSpH 7,4, 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: GluN2A/NR2A
Clonality: monoclonal
Clone: S327A-38
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat