Order Entry
Export
ContactUsLinkComponent
Anti-SLC1A1 Mouse Monoclonal Antibody [clone: S180-41]
Anti-SLC1A1 Mouse Monoclonal Antibody [clone: S180-41]
Catalog # USBI167584
Supplier:  US Biological
Anti-SLC1A1 Mouse Monoclonal Antibody [clone: S180-41]
Catalog # USBI167584
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    neuronal/epithelial high affinity glutamate transporter, system Xag, member 1
  • Antigen symbol:
    Solute carrier family 1, member 1
  • Clonality:
    Monoclonal
  • Clone:
    S180-41
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage buffer:
    PBSpH 7,4, 50% glycerol.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: 5 ?g

Type: Primary
Antigen: SLC1A1 (solute carrier family 1 (neuronal/epithelial high affinity glutamate transporter, system Xag), member 1)
Clonality: monoclonal
Clone: S180-41
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human