Order Entry
Export
ContactUsLinkComponent
Anti-Tropomyosin Mouse Monoclonal Antibody [clone: TM-36]
Anti-Tropomyosin Mouse Monoclonal Antibody [clone: TM-36]
Catalog # USBI145323
Supplier:  US Biological
Anti-Tropomyosin Mouse Monoclonal Antibody [clone: TM-36]
Catalog # USBI145323
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Tropomyosin
  • Clonality:
    Monoclonal
  • Clone:
    TM-36
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Rabbit,
    Chicken,
    Human
  • Western blot:
    Yes
  • Format:
    ascites
  • Form:
    lyophilized powder
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: Tropomyosin
Clonality: monoclonal
Clone: TM-36
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Chicken, Human, Rabbit