Anti-Phosphoserine Mouse Monoclonal Antibody [clone: PS-53]
USBI145053
- Antibody type:Primary
- Antigen name:Phosphoserine
- Antigen symbol:pS
- Clonality:Monoclonal
- Clone:PS-53
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Chicken
- Western blot:Yes
- Format:ascites
- Form:lyophilized powder
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: Phosphoserine
Clonality: monoclonal
Clone: PS-53
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken