Order Entry
Export
ContactUsLinkComponent
 
Anti-p19INK4d Mouse Monoclonal Antibody [clone: IMD-19]
Catalog # USBI145020
Supplier:  US Biological
Anti-p19INK4d Mouse Monoclonal Antibody [clone: IMD-19]
Catalog # USBI145020
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    P19INK4D
  • Clonality:
    Monoclonal
  • Clone:
    IMD-19
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Format:
    ascites
  • Form:
    lyophilized powder
  • Storage temperature:
    Cool
  • Concentration:
    ~0,1 mg/ml (after reconstitution)
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: p19INK4d
Clonality: monoclonal
Clone: IMD-19
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human