Anti-NCAM1 Mouse Monoclonal Antibody [clone: IML-43]
Catalog # USBI144947
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Neural Cell Adhesion Molecule 1
- Antigen symbol:NCAM1
- Clonality:Monoclonal
- Clone:IML-43
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat
- Western blot:Yes
- Format:ascites
- Form:lyophilized powder
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: NCAM1 (neural cell adhesion molecule 1)
Clonality: monoclonal
Clone: IML-43
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat