Order Entry
Export
ContactUsLinkComponent
Anti-IL-17/IL-17A Rabbit Polyclonal Antibody
Anti-IL-17/IL-17A Rabbit Polyclonal Antibody
Catalog # USBI144734
Supplier:  US Biological
Anti-IL-17/IL-17A Rabbit Polyclonal Antibody
Catalog # USBI144734
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 17
  • Antigen symbol:
    IL17
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Form:
    lyophilized powder
  • Antigen synonyms:
    IL-17A
  • Storage buffer:
    0,9mg NaCl, 0,2mg Na2HPO4, 0,05mg NaN3.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: IL-17/IL-17A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse