Anti-Interleukin 12 Rat Monoclonal Antibody
USBI143603
- Antibody type:Primary
- Antigen name:Interleukin 12
- Antigen symbol:IL12
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- Isotype:IgG1 kappa
- Reactivity:Mouse
- Western blot:Yes
- Form:lyophilized powder
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:500 µg
- Pk:500 µG
Recommended dilution: ELISA: 4-8?g/ml
Type: Primary
Antigen: Interleukin 12
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1 Kappa
Reactivity: Mouse