Order Entry
Export
ContactUsLinkComponent
Anti-TFPI2 Mouse Monoclonal Antibody [clone: 13k101]
Anti-TFPI2 Mouse Monoclonal Antibody [clone: 13k101]
  USBI143228
undefined
Anti-TFPI2 Mouse Monoclonal Antibody [clone: 13k101]
  USBI143228

 

  • Antibody type:
    Primary
  • Antigen name:
    Tissue Factor Pathway Inhibitor 2
  • Antigen symbol:
    TFPI2
  • Clonality:
    Monoclonal
  • Clone:
    13k101
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Format:
    ascites
  • Form:
    liquid
  • Storage buffer:
    ascites fluid.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: TFPI2 (tissue factor pathway inhibitor 2)
Clonality: monoclonal
Clone: 13k101
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human