Anti-DEFA5 Mouse Monoclonal Antibody [clone: 13k18]
Catalog # USBI143055
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:defensin, alpha 5, Paneth cell-specific
- Antigen symbol:DEFA5
- Clonality:Monoclonal
- Clone:13k18
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Amino acid number:16 to 31
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: DEFA5 (defensin, alpha 5, Paneth cell-specific)
Clonality: monoclonal
Clone: 13k18
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse