Anti-Fetoprotein Goat Polyclonal Antibody
Catalog # USBI137958
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Fetoprotein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:affinity-purified
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: Fetoprotein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: