Anti-CA 19-9 Mouse Monoclonal Antibody [clone: 13H12]
Catalog # USBI137580
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:CA 19-9
- Clonality:Monoclonal
- Clone:13H12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgM
- Form:liquid
- Storage buffer:10mMTRIS, pH 7,5, 150mM sodium chloride, 0,05% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Primary
Antigen: CA 19-9
Clonality: monoclonal
Clone: 13H12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: