Anti-CA 19-9 Mouse Monoclonal Antibody [clone: 13H10]
USBI137578
- Antibody type:Primary
- Antigen symbol:CA 19-9
- Clonality:Monoclonal
- Clone:13H10
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: CA 19-9
Clonality: monoclonal
Clone: 13H10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human