Anti-DARPP 32 Rabbit Monoclonal Antibody [clone: 13G27]
Catalog # USBI137428
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:DARPP 32
- Clonality:Monoclonal
- Clone:13G27
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Modification:phospho-Thr34
- Storage buffer:10mMsodium HEPES, pH 7,5, 150mM NaCl, 100ug/ml BSA, 50% glycerol and <0,02% sodium azide.
- Sequence:SyntheticphosphopeptidecorrespondingtoresiduessurroundingT34ofhumanDARPP32SpeciesSequenceHomology:mouserat
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: DARPP 32
Clonality: monoclonal
Clone: 13G27
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human