Anti-VAMP 1,2 Mouse Monoclonal Antibody [clone: SP-11]
Catalog # USBI136854
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:VAMP 1,2
- Clonality:Monoclonal
- Clone:SP-11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 50% glycerol.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry: parrafin
Type: Primary
Antigen: VAMP 1,2
Clonality: monoclonal
Clone: SP-11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human