Anti-Mucin 13 Rabbit Polyclonal Antibody
Catalog # USBI136269
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Mucin 13
- Antigen symbol:MUC13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Form:liquid
- Storage buffer:0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 10-15 ?g/ml
Type: Primary
Antigen: Mucin 13
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human