Order Entry
Export
ContactUsLinkComponent
Anti-KIAA1324 Rabbit Polyclonal Antibody
Anti-KIAA1324 Rabbit Polyclonal Antibody
Catalog # USBI136209
Supplier:  US Biological
Anti-KIAA1324 Rabbit Polyclonal Antibody
Catalog # USBI136209
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    KIAA1324
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Immunochemistry: 10-15 ?g/ml

Type: Primary
Antigen: KIAA1324 (KIAA1324)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human