Anti-ZNF268 Mouse Monoclonal Antibody [clone: 3B4]
USBI135667
- Antibody type:Primary
- Antigen name:zinc finger protein 268
- Antigen symbol:ZNF268
- Clonality:Monoclonal
- Clone:3B4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:liquid
- Amino acid number:1 to 90
- Storage buffer:PBS, pH 7,2.
- Sequence:MATRVRTASIWVPPLQERNSSWDRIRKLQGQESILGQGTPGLQPLPGTPRQKQKSRRIEKVLEWLFISQEQPKITKSWGPLSFMDVFVDF
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: ZNF268 (zinc finger protein 268)
Clonality: monoclonal
Clone: 3B4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human